QuikChange II
Lieferant: STRATAGENE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
200555 200521 200522 200523 200524 200555
STRA200555EA 0 EUR InStock
STRA200555 AGLS200521 AGLS200522 AGLS200523 AGLS200524 AGLS200555
QuikChange II
Nukleinsäure-Reagenzien Modifikation und Markierung von Nukleinsäure
These QuikChange II kits provide improved fidelity, while maintaining greater than 80% mutation efficiency for single site mutagenesis.
  • Includes PfuUltra High-fidelity DNA polymerase to minimise unwanted errors
  • Complete kit provides all reagents necessary to support mutagenesis (at single sites)
Used for large constructs, and for use with electroporation competent cells.
Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge