Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Site-directed mutagenesis kits, QuikChange Lightning

Lieferant: AGILENT

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


210515 210516 210518 210519 210513 210514
AGLS210515EA 551 EUR InStock
AGLS210515 AGLS210516 AGLS210518 AGLS210519 AGLS210513 AGLS210514
Site-directed mutagenesis kits, QuikChange Lightning
Nukleinsäure-Reagenzien Modifikation und Markierung von Nukleinsäure
The fastest and latest generation of the market-leading QuikChange kits speeds up the protocol for performing single and multiple site-directed mutagenesis to less than 3 hours.

  • For single site mutagenesis, one kit accommodates both short and long templates (4 to 14 kB)
  • >80% mutagenesis efficiency for 1 site
  • >55% mutagenesis efficiency for 3 mutations simultaneously

These QuikChange Lightning kits, like their predecessors, are based on a linear amplification method whereby mutation-containing primers are incorporated but not copied.

The protocols of other commercially available kits rely on PCR-based methods, which may result in unwanted second-site errors. The QuikChange Lightning Kit allows you to introduce point mutations, insertions, or deletions twice as fast compared to earlier kit versions, and the QuikChange Lightning Multi Kit delivers mutants up to three times faster than our original multi-site mutagenesis kit, both kits accomplish this without sacrificing the mutagenesis efficiency, reliability and accuracy.

Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.


Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge