Benchtop cooler, cryo preservation module, StrataCooler, AGLS
Lieferant: AGILENT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
400005
AGLS400005EA 314 EUR InStock
AGLS400005
Benchtop cooler, cryo preservation module, StrataCooler, AGLS
Kühlbehälter Tischkühler
Most cell types show >50% survival when cooling rates are kept at 0,2 to 1 °C/min. The StrataCooler Cryo preservation module has been designed to freeze mammalian and insect cells at a controlled rate of 0,4 to 0,6 °C/min. to achieve an 80 to 90% survival rate.
  • 32-(standard) cryovial capacity
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge