Master mixes, brilliant HRM ultra-fast loci
Lieferant: AGILENT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
5190-7827 5190-7909 600536
AGLS5190-7827EA 205 EUR InStock
AGLS5190-7827 AGLS5190-7909 AGLS600536
Master mixes, brilliant HRM ultra-fast loci
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzyme und Kits
The Brilliant HRM Ultra-Fast Loci Master Mix for High Resolution Melt (HRM) analysis.
  • MgCl₂ and dNTPs are included at optimised concentrations
  • Master Mix is stable after multiple freeze thaws, reducing wastage and increasing batch-to-batch reliability
  • Start Taq polymerase enzyme requires only three minutes for activation
  • The dye is non-toxic and can be added at saturating concentrations - insuring minimal inhibition while preserving high sensitivity
  • Master Mix can be used with the AriaMx Real-Time PCR System or any HRM-capable thermal cycler
The Master Mix combines a mutant Fast-Start Taq polymerase, optimised concentrations of MgCl₂ and dNTPs, and EvaGreen, release-on-demand dye to enable researchers to mix-and-go faster and with total confidence - even for the most difficult genotypes.
The optimised MgCl₂ and dNTP concentrations reduce assay preparation time, while low surfactant levels minimise bubble trouble during pipetting. In addition, the 12-month storage at ‒20 °C is more than double that of other HRM reagents.
The passive reference dye may be added to fluorescence-based quantitative PCR assays to compensate for non-PCR related variations in fluorescence.
Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge