Porcine detection kit
Lieferant: AGILENT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
5500-0094
AGLS5500-0094EA 1472 EUR InStock
AGLS5500-0094
Porcine detection kit
Nukleinsäure-Reagenzien Enzyme und Kits für Endpunkt-PCR
Detect as low as 300 fg of Porcine DNA in food and pharmaceutical samples, including gel caps.
  • Multiplex detection of porcine specific DNA and Alien DNA control
  • DNA Isolation Module, with spin cup protocol
  • qPCR assay to detect nucleic acids of Porcine origin
  • No interference from Alien assay
The Agilent Porcine Detection Kit has two components: a DNA Isolation Module and a qPCR Kit. The Porcine Detection Kit contains reagents and materials for the extraction and isolation of DNA from food samples and other materials, including gel caps, as well as the subsequent qPCR amplification and detection of porcine DNA down to 300 fg.
Because isolated DNA may contain contaminants that inhibit PCR, the qPCR kit amplifies and detects two distinct targets in a single reaction: a porcine-specific DNA sequence and an external DNA control that enables detection of PCR inhibition.
Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge