Femto Pulse 12-Capillary Array
Lieferant: AGILENT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A1600-12502240
AGLSA1600-12502240EA 2940 EUR InStock
AGLSA1600-12502240
Femto Pulse 12-Capillary Array
Elektrophorese-Systeme Kapillarelektrophorese-Systeme
12-Capillary Array for analysis on the Femto Pulse system.
  • High resolution - optimised length for enhanced resolution; Run Multiple Samples - Run up to 12 samples at same time
  • Low sensitivity - integral part of the Femto Pulse system aiding in low sensitivity detection
The Femto Pulse 12-Capillary Array is used with the Genomic DNA 165 kb kit, the Ultra Sensitivity NGS kit (1 to 6000 bp), the Ultra Sensitivity RNA kit, and the 55 kb BAC kit. It has a separation or effective length of 22 cm, with an overall length of 40 cm. The 12 parallel capillary array is ideal for lower throughput laboratories.
Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge