Small RNA Kits
Lieferant: AGILENT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
DNF-470-0275
AGLSDNF-470-0275EA 1280 EUR InStock
AGLSDNF-470-0275
Small RNA Kits
Elektrophorese-Systeme Kapillarelektrophorese-Systeme
Analysis of small RNA quality control including microRNA quantification.
  • Broad concentration range - The small RNA kit has a concentration range of 25 to 2500 pg/µl
  • Short analysis time - Analysis time in as little as 22 minutes
  • Reliable sizing - Sizing range from 15 to 200 nt ensures accurate and precise sizing of small RNA samples
  • Low sample volume - Only 2 µl of sample is required, minimising sample loss to QC steps
  • Specific RNA analysis modes - ProSize data analysis software uses a specific small RNA analysis region function automatically calculating percent microRNA and quantification of microRNA and small RNA
  • Capillary array - Optimised for the 12 -capillary array
The small RNA kit focuses on microRNA and small RNA quality control. Analysis of small RNAs is essential for downstream applications such as small RNA NGS libraries. The small RNA ladder includes six fragments ranging from 15 to 200 nt, which provides accurate and precise quantification and sizing of small RNA and microRNA. Focusing on a narrow range of 200 nt magnifies the individual areas of microRNA (10 to 40 nt) and small RNA (40 to 200 nt) allowing for high separation resolution and detailed analysis of each region.
Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge