Anti-GOGAT Rabbit Polyclonal Antibody
Lieferant: Agrisera
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AS07242
AGRIAS07242EA 329 EUR InStock
AGRIAS07242
Anti-GOGAT Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GOGAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Miscanthus giganteus, Phaseolus vulgaris, Spartina sp., Zea mays
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge