Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 594)
Lieferant: Agrisera
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AS122056
AGRIAS122056EA 317 EUR InStock
AGRIAS122056
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 594)
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 594
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Horse, Human, Mouse, Pig, Rabbit
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge