Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-UAGPase Rabbit Polyclonal Antibody

Lieferant: Agrisera

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

AS142829
AGRIAS142829EA 329 EUR InStock
AGRIAS142829
Anti-UAGPase Rabbit Polyclonal Antibody
Antikörper
UAGPase (UDP-GlcNAc pyrophosphorylase) is involved in the biosynthesis of UDP-glucosamine, an essential precursor for glycoprotein and glycolipid synthesis and is also used for regulatory protein modification in signaling pathways. The enzyme is localized in a cytoplasm. Alternative names: N-acetylglucosamine-1-phosphate uridylyltransferase 2, UDP-N-acetylgalactosamine diphosphorylase 2, UTP--glucose-1-phosphate uridylyltransferase 2, AGX.,UAGPase (UDP-GlcNAc pyrophosphorylase) is involved in the biosynthesis of UDP-glucosamine, an essential precursor for glycoprotein and glycolipid synthesis and is also used for regulatory protein modification in signaling pathways. The enzyme is localized in a cytoplasm. Alternative names: N-acetylglucosamine-1-phosphate uridylyltransferase 2, UDP-N-acetylgalactosamine diphosphorylase 2, UTP--glucose-1-phosphate uridylyltransferase 2, AGX.

Type: Primary
Antigen: UAGPase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, H. vulgare, N. tabacum

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge