Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MIP1 Rabbit Polyclonal Antibody

Lieferant: Agrisera

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

AS152826
AGRIAS152826EA 329 EUR InStock
AGRIAS152826
Anti-MIP1 Rabbit Polyclonal Antibody
Antikörper
MIP1 (Aquaporin, glycerol transport activity) is localized to the contractile vacuole, which in most freshwater flagellates is used to expel excess water.,MIP1 (Aquaporin, glycerol transport activity) is localized to the contractile vacuole, which in most freshwater flagellates is used to expel excess water.
Reacts with Chlamydomonas reinhardtii (strains CC3395, and UVM4 from Neupert et al., 2009. (GeneRation of Chlamydomonas Strains that Efficiently Express Nuclear Transgenes.)

Type: Primary
Antigen: MIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge