Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-6x His Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: AD1.1.10]

Lieferant: Agrisera

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

AS152930
AGRIAS152930EA 482 EUR InStock
AGRIAS152930
Anti-6x His Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: AD1.1.10]
Antikörper
Mouse anti-6x Histidine is a primary antibody which binds to 6x Histidine tag and is directly conjugated to HRP. This is of advantage to shorten assay time by no need to use a secondary antibody to 6x Histidine tag. ,Mouse anti-6x Histidine is a primary antibody which binds to 6x Histidine tag and is directly conjugated to HRP. This is of advantage to shorten assay time by no need to use a secondary antibody to 6x Histidine tag.

Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: AD1.1.10
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge