Anti-Tubulin Alpha Chain Mouse Monoclonal Antibody [clone: TU-01] (FITC)
Lieferant: Agrisera
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AS214626
AGRIAS214626EA 282 EUR InStock
AGRIAS214626
Anti-Tubulin Alpha Chain Mouse Monoclonal Antibody [clone: TU-01] (FITC)
Antikörper
Recommended dilution: 1 to 4 µg/ml (Flowcytometry)
Type:
Antigen: Tubulin Alpha Chain
Clonality: Monoclonal
Clone: TU-01
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Arabidopsis thaliana, Eisenia, Nicotiana tabacum, Paramecium, Saccharomyces cerevisiae, human, mouse, pig, turkey
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge