Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PIP2;1, PIP2;2, PIP2;3 Rabbit Polyclonal Antibody

Lieferant: Agrisera

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

AS224810
AGRIAS224810EA 230 EUR InStock
AGRIAS224810
Anti-PIP2;1, PIP2;2, PIP2;3 Rabbit Polyclonal Antibody
Antikörper
PIP2;2 is a plasma membrane aquaporin.

  • For reconstitution add 50 µl, of sterile or deionised water

Spin tubes briefly prior to opening them to avoid any losses that might occur from lyophilised material adhering to the cap or sides of the tubes.

Recommended dilution: 1:1000 (WB).

Molecular weight: 3,4 kDa (due to N-terminal or C-terminal processing).

Achtung! Bitte beachten: For research use only.

Type: Primary
Antigen: PIP2;1, PIP2;2, PIP2;3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Arabidopsis thaliana

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge