Anti-PetL Rabbit Polyclonal Antibody
Lieferant: Agrisera
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AS224883
AGRIAS224883EA 329 EUR InStock
AGRIAS224883
Anti-PetL Rabbit Polyclonal Antibody
Antikörper
PetL (Cytochrome b6-f complex subunit 6) is a part of the cytochrome b6-f complex, which mediates electron transfer between photosystem II (PSII) and photosystem I (PSI), cyclic electron flow around PSI, and state transitions. The protein is of importance for photoautotrophic growth as well as for electron transfer efficiency and stability of the cytochrome b6-f complex.
  • For reconstitution add 50 µl, of sterile or deionised water
Spin tubes briefly prior to opening them to avoid any losses that might occur from lyophilised material adhering to the cap or sides of the tubes.
Recommended dilution: 1:1000 (WB).
Molecular weight: 3,4 kDa (due to N-terminal or C-terminal processing).
Achtung! Bitte beachten: For research use only.
Type: Primary
Antigen: PetL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Nicotiana tabacum
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge