Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-UGPase Rabbit Polyclonal Antibody

Lieferant: Agrisera

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

AS234916
AGRIAS234916EA 329 EUR InStock
AGRIAS234916
Anti-UGPase Rabbit Polyclonal Antibody
Antikörper
UDP-glucose pyrophosphorylase (UGPase, UDPGP) is a key enzyme of synthesis of sucrose, cellulose and other saccharides. There are two cytoplasmic isoforms of UGPase-A (which share 94% identity on amino acid level) and one chloroplastic UGPase-B isoform in Arabidopsis thaliana which share ca. 10 to 11% of identity (Kleczkowski et al. 2011).

  • For reconstitution add 50 µl, of sterile or deionised water

Spin tubes briefly prior to opening them to avoid any losses that might occur from lyophilised material adhering to the cap or sides of the tubes.

Recommended dilution: 1:1000 to 1:5000 (WB).

Expected molecular weight: 51,7 kDa
Apparent molecular weight: 52,3 kDa

Achtung! Bitte beachten: For research use only.

Type: Primary
Antigen: UGPase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Arabidopsis thaliana

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge