Bovine serum albumin (BSA)
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ADI-80-1928
ALEXADI-80-1928EA 104 EUR InStock
ALEXADI-80-1928
Bovine serum albumin (BSA)
Proteine und Peptide
This BSA Solution (10%) is specially designed to create the Blocking Buffer and Assay Buffer in ImmunoSet products. Each bottle is sufficient for use with 5×96 wells microtiter plates if following the instructions in our ImmunoSet protocol. The BSA Solution may be used for any other assay development needs. Additional assay development reagents are available as companion products to our ImmunoSet products.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge