Matriptase (human), ELISA kit
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ADI9002210001
ALEXADI9002210001EA 957 EUR InStock
ALEXADI9002210001
Matriptase (human), ELISA kit
Tests ELISAs
Sensitive, rapid ELISA kit to quantify matriptase, a serine protease and unique cancer biomarker.
For the quantitative determination of human matriptase in plasma and serum.
The Matriptase ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of matriptase in plasma and serum with results in just 3 hours.
Bestellinformation: Contains Microtiter Plate, Standards, Calibrators, Antibody-conjugate, Streptavidin-HRP, TMB Reagent, Stop Solution, Wash Buffer Concentrate.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge