Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Fusicoccin ≥90% (durch HPLC), weißer Feststoff

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


BML-EI334-0001
ALEXBML-EI334-0001EA 344 EUR InStock
ALEXBML-EI334-0001
Fusicoccin ≥90% (durch HPLC), weißer Feststoff
Fusicoccin
Plant H+-ATPase activator.

Diterpene glucoside with numerous effects. Induces H+ extrusion from plant cells by 14-3-3-dependent activation of the plasma membrane H+-ATPase. It induces translocation of 14-3-3 proteins from the cytosol to the membrane and causes a dramatic increase in cytosolic 14-3-3s. Stimulates release of cytochrome c from mitochondria, cell enlargement, ion uptake, seed germination, opening the stomata, etc. Markedly reduces the cell's capability for H2O2 scavenging. Fusicoccin has been demonstrated to cause randomization of asymmetry of the left-right axis (heterotaxia) in Xenopus embryos.

Isolated from fungus Fusicoccum amygdali.

Soluble in ethanol and chloroform. Slightly soluble in ethyl acetate, ethyl ether, sparingly soluble in acetone.

Achtung! Bitte beachten: Stable for at least 1 year after receipt when stored, as supplied, at -20 °C. Stock solutions are stable for up to 3 months at -20 °C.


Formel: C₃₆H₅₆O₁₂
MW: 680.8 g/mol
MDL Number: MFCD00070322
CAS-Nummer: 20108-30-9
Order Now
Spezifikation Testergebnisse
Purity ≥90% (HPLC)

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge