Pipette tips
Lieferant: ALPHA LABORATORIESCHEMICALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
LW6470 LW6365S LW6393S LW1154 LW6361 LW6471 LW1126 LW1136 LW6391 LW6360 LW6475S
ALPCLW6470EA 112 EUR InStock
ALPCLW6470 ALPCLW6365S ALPCLW6393S ALPCLW1154 ALPCLW6361 ALPCLW6471 ALPCLW1126 ALPCLW1136 ALPCLW6391 ALPCLW6360 ALPCLW6475S
Pipette tips
Pipettenspitzen
These tips are manufactured from medical grade PP in modern production facilities with stringent quality controls at every stage.
  • Tips are lot tested and certified for absence of detectable human DNA and detectable endotoxins (pyrogens)
  • Tips are tested and certified for absence of heavy metals, detectable DNase and RNase
  • With graduations to visually check volume
  • Suitable for all major pipettor brands
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge