ASTM® precision thermometers, low hazard
Lieferant: AMARELL, ARNO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A300770-CB/CC4 A318188CC5 A318192CC5 A318132CC3 A318196CC5 A318200CC5 A318204CC5
AMARA300770-CB/CC4EA 0 EUR InStock
AMARA300770-CB/CC4 AMARA318188CC5 AMARA318192CC5 AMARA318132CC3 AMARA318196CC5 AMARA318200CC5 AMARA318204CC5
ASTM® precision thermometers, low hazard
Thermometer Analoge Thermometer
Low hazard precision thermometers are used for special applications and suitable for government calibration.
  • Solid stem and white backed
  • Round or prismatic with round top finish
  • Durable pigment
  • Non-wetting because of capillary tube specially coated inside
Zertifizierungen: ASTM standard E2251–07 (Standard specification for liquid-in-glass ASTM thermometers with low-hazard precision liquids).
Verpackung: Packaged in transparent square plastic case.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge