Digital thermometer, Maxi Pen
Lieferant: AMARELL, ARNO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
E905190
AMARE905190EA 32.92 EUR InStock
AMARE905190
Digital thermometer, Maxi Pen
Thermometer Digitalthermometer
Resistant and intensive thermometer for fast temperature indication. With fast-responding stainless steel penetration probe (L×Ø: 125×3,5 mm).
  • 'HOLD' feature
  • Automatic power-off after 10 min.
  • °C / °F switchable
  • Waterproof casing
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge