Digital thermometer, DE 305
Lieferant: AMARELL, ARNO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
E908650
AMARE908650EA 148.36 EUR InStock
AMARE908650
Digital thermometer, DE 305
Thermometer Digitalthermometer
Universal type-K thermometer for interchangeable thermo-couple probes for measurements in high temperature ranges.
  • 'Hold' function
  • Maximum value storing
  • Calibration function
Features like gripping and robust rubber protective sheath in extent of supply.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge