Electronic precision thermometer for foodstuff
Lieferant: AMARELL, ARNO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
E910600
AMARE910600EA 0 EUR InStock
AMARE910600
Electronic precision thermometer for foodstuff
Thermometer Digitalthermometer
Digital thermometer type ad 17th is a piercing thermometer, very well suitable for core measurements on roasting and freezing.
  • Wide measuring range
  • Precise PT100 probe of stainless steel
  • Handle of probe and PTFE cable resistant up to 250 °C
Achtung! Bitte beachten: or 0,5 % of range
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge