Pocket thermometers, with clip
Lieferant: AMARELL, ARNO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
G15602-FL G15606-FL
AMARG15602-FLEA 36.68 EUR InStock
AMARG15602-FL AMARG15606-FL
Pocket thermometers, with clip
Thermometer Analoge Thermometer
The thermometers with Hg filling have a blue reflecting capillary.
  • Pen size
  • Total immersion
  • Enclosed scale
Lieferumfang: All pocket thermometers supplied in nickel plated double receptacle.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge