Density hydrometers, S50
Lieferant: AMARELL, ARNO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H811108 H811100 H811102 H811104 H811106 H811110 H811112 H811114 H811116 H811118 H811120 H811122 H811124 H811126 H811128 H811130 H811132 H811134 H811136 H811138 H811140 H811142 H811144 H811146 H811148 H811150 H811152 H811154
AMARH811108EA 50.82 EUR InStock
AMARH811108 AMARH811100 AMARH811102 AMARH811104 AMARH811106 AMARH811110 AMARH811112 AMARH811114 AMARH811116 AMARH811118 AMARH811120 AMARH811122 AMARH811124 AMARH811126 AMARH811128 AMARH811130 AMARH811132 AMARH811134 AMARH811136 AMARH811138 AMARH811140 AMARH811142 AMARH811144 AMARH811146 AMARH811148 AMARH811150 AMARH811152 AMARH811154
Density hydrometers, S50
Aräometer
Hydrometers in a wide range of measuring scales.
  • Shot weighted body
  • Paper scale with colour code and type designation
Suitable for government verification.
Zertifizierungen: DIN 12791
Verpackung: In a transparent plastic case.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge