Industrial thermometers, straight stem
Lieferant: AMARELL, ARNO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
T90004-FL T90128-FL T89950-FL T89952-FL T89954-FL T89958-FL T90000-FL T90008-FL T90012-FL T90016-FL T90020-FL T90024-FL T90032-FL T90036-FL T90052-FL T90056-FL T90060-FL T90064-FL T90068-FL T90072-FL T90076-FL T90084-FL T90104-FL T90108-FL T90112-FL T90116-FL T90120-FL T90124-FL T90136-FL T90160-EF T90164-EF T90168-EF T90176-EF T90180-EF T90212-GA T90220-GA T90228-GA T90232-GA
AMART90004-FLEA 37.09 EUR InStock
AMART90004-FL AMART90128-FL AMART89950-FL AMART89952-FL AMART89954-FL AMART89958-FL AMART90000-FL AMART90008-FL AMART90012-FL AMART90016-FL AMART90020-FL AMART90024-FL AMART90032-FL AMART90036-FL AMART90052-FL AMART90056-FL AMART90060-FL AMART90064-FL AMART90068-FL AMART90072-FL AMART90076-FL AMART90084-FL AMART90104-FL AMART90108-FL AMART90112-FL AMART90116-FL AMART90120-FL AMART90124-FL AMART90136-FL AMART90160-EF AMART90164-EF AMART90168-EF AMART90176-EF AMART90180-EF AMART90212-GA AMART90220-GA AMART90228-GA AMART90232-GA
Industrial thermometers, straight stem
Thermometer Analoge Thermometer
Capillary prismatic thermometers.
  • Yellow backed
  • Partial immersion
  • Milk glass scale
Cork-glass top finish.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge