Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Surgical Gloves, Latex, ENCORE® Latex Textured

Lieferant: ANSELL MEDICAL PRODUCTS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

330101055 330101060 330101065 330101070 330101075 330101080 330101085 330101090
ANSM330101055EA 76.8 EUR InStock
ANSM330101055 ANSM330101060 ANSM330101065 ANSM330101070 ANSM330101075 ANSM330101080 ANSM330101085 ANSM330101090
Surgical Gloves, Latex, ENCORE® Latex Textured
Handschuhe
Latex, powder-free surgical gloves ensuring secure grip for confident instrument handling.

  • Textured finish: Secure grip of instruments under dry or damp conditions
  • Optimised thickness: Ideal balance of tactile sensitivity and protection
  • Proprietary donning technology: Effortless donning and doffing with either dry or damp hands

Enhanced grip: The textured finish of these sterile latex gloves delivers exceptional grip of medical instruments under dry or damp conditions, making them ideal outer gloves when double gloving.
Heightened tactile sensitivity: The unique latex formulation and processes enable the thickness of these sterile gloves to be optimised for the wearer to experience the ideal balance of tactile sensitivity and hand protection.

Effortless donning and doffing: ENCORE® Latex Textured surgical gloves are designed with a proprietary internal coating to facilitate easy dry and damp donning when first worn or during intra-operative changes.
Chemo-tested: Tested to ASTM D6978, these textured latex gloves are safe to use with a wide range of chemotherapy drugs.

Recommended for:
Enhanced grip in wet environments
Extra comfort during long, sustained procedures
Damp donning
Use as an outer glove when double gloving

Zertifizierungen: ASTM D3577, ASTM D6978, ASTM D7160, ASTM F739, EN 16523-1, EN 374-2 and -4, EN 455 1-4,
EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, ISO 10282.

Verpackung: 50 pairs per box; 4 boxes per carton/case; 200 pairs per carton/case.

Achtung! Bitte beachten: Safe use of these gloves by latex-sensitised individuals has not been established. This product contains natural rubber latex, which may cause allergic reactions.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge