Apoptosis inducer set
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
API-16S
API-16SEA 1360 EUR InStock
API-16S
Apoptosis inducer set
Tests Zelltests
Apoptosis inducers are used for induction of apoptosis in cultured cells. The selection of apoptosis inducers include actinomycin D, camptothecin, cycloheximide, dexamethasone, doxorubicin and etoposide.
  • Ready to use
  • Long storage life
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb340ab59b78aa142431268e23ef95b493013f5b&vid=fb340ab59b78aa142431268e23ef95b493013f5b&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2aa4423dbded4a11a8b76daa453019131515b275&vid=2aa4423dbded4a11a8b76daa453019131515b275&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34ad33574342ded20e1a2a1e4a4d1a62753f0721&vid=34ad33574342ded20e1a2a1e4a4d1a62753f0721&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d973f8a38d16090d3bbba7e7b241cd7d80b6902&vid=3d973f8a38d16090d3bbba7e7b241cd7d80b6902&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c5d5356c4f5ab01f84afd6c0417d07d7a8c330d&vid=8c5d5356c4f5ab01f84afd6c0417d07d7a8c330d&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge