Poly(caprolactone-co-glycolide) 90:10
Lieferant: Apollo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BIBE1003-5G
APOSBIBE1003-5GEA 63.8 EUR InStock
APOSBIBE1003-5G
Poly(caprolactone-co-glycolide) 90:10
Poly(caprolactone-co-glycolide)
Polymer. Caprolactone:glycolide 90:10. Viscosity 0.75 dL/g
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge