BICL207-100MG
APOSBICL207-100MGEA
132
EUR
InStock
APOSBICL207-100MG
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)
Primary amine & Sulphhydryl reactiveEnzyme-antibody conjugation -Stable maleimide groupRef: Hushida, S. et al (1984) J.Applied Biochem. 56,56-63