Acetyl pepstatin
Lieferant: Apollo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BIMI7332-25MG
APOSBIMI7332-25MGEA 40.6 EUR InStock
APOSBIMI7332-25MG
Acetyl pepstatin
Proteine und Peptide
Inhibitor of aspartyl proteases, particularly effective onHIV-I proteinase and HIV-II proteinase
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge