Sulpho-SAND (Sulphosuccinimidyl 2-(3-azido-2-nitrobenzamido)ethyl-1,3'-dithiopropionate)
Lieferant: Apollo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BIPA111-100MG
APOSBIPA111-100MGEA 174 EUR InStock
APOSBIPA111-100MG
Sulpho-SAND (Sulphosuccinimidyl 2-(3-azido-2-nitrobenzamido)ethyl-1,3'-dithiopropionate)
Sulpho-SAND (Sulphosuccinimidyl 2-(3-azido-2-nitrobenzamido)ethyl-1,3'-dithiopropionate)
Photoreactive -Primary amine reactive Thiol cleavable -Water soluble
Ref: Lewis,R.V,et al(1977) Biochemistry 16, 5650-5654
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge