Banana powder
Lieferant: APOLLO SCIENTIFIC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BIPB852-100G
APOSBIPB852-100GEA 21.9 EUR InStock
APOSBIPB852-100G
Banana powder
Gastronomiezubehör
A mixture of natural banana puree and maltodextrin. Use at a concentration of 30 to 50 g/L.
  • MFCD00130601
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge