Pentostatin 99%
Lieferant: Apollo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BISN0008-5MG
APOSBISN0008-5MGEA 43.5 EUR InStock
APOSBISN0008-5MG
Pentostatin 99%
Pentostatin
Pentostatin can inhibit adenosine deaminase irreversibly with Ki of 2.5 pM, as an antineoplastic anti-metabolite used in the treatment of several forms of leukemia.
Formel: C₁₁H₁₆N₄O₄
Storage Temperature: Gefrierschrank
MDL Number: MFCD00078802
CAS-Nummer: 53910-25-1
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge