OR52255-100G
OR52255-25G
OR52255-1G
OR52255-5G
APOSOR52255-100GEA
48
EUR
InStock
APOSOR52255-100G
APOSOR52255-25G
APOSOR52255-1G
APOSOR52255-5G
2-Chlorphenol 99%
2-Chlorphenol
Has been used as a reagent for the protection of OH-groups in oligonucleotide synthesis. A study of various cleavage methods showed that the tetramethylguanidine salts of either 4-nitrobenzaldoxime or pyridine-2-carboxaldoxime gave the best results: Tetrahedron Lett., 2727 (1978).