3-[(Methylsulphonyl)thio]propanoic acid
Lieferant: Apollo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
Achtung
OR7810T-100MG
APOSOR7810T-100MGEA 284 EUR InStock
APOSOR7810T-100MG
3-[(Methylsulphonyl)thio]propanoic acid
3-[(Methylsulphonyl)thio]propanoic acid
Reacts specifically and rapidly with thiols to form mixed disulfides. Useful for probing the structures & functions of receptor & ion channel proteins.
Formel: C₄H₈O₄S₂
MW: 184.234 g/mol
Melting Pt: 60…62 °C
Storage Temperature: Gefrierschrank
MDL Number: MFCD00797599
CAS-Nummer: 92953-12-3
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge