Murashige and skoog modified basal salts (½ Micros and ½ Macros)
Lieferant: Apollo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PMM153-50L
APOSPMM153-50LEA 0 EUR InStock
APOSPMM153-50L
Murashige and skoog modified basal salts (½ Micros and ½ Macros)
Zellkulturmedien
Contains macro/micronutrients, sucrose, agar and organics as described by Murashige and Skoog, 1962 Plant tissue culture testedFull specification given in appendix.
  • MFCD00240976
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge