Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Methanol ≥99,9% (durch GC) Reag. Ph. Eur., ACS, ISO zur Analyse

Lieferant: APPLICHEM

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

131091.1212 131091.0716 131091.1211 131091.1214 131091.1611 131091.1612
APPC131091.1212EA 69.3 EUR InStock
APPC131091.1212 APPC131091.0716 APPC131091.1211 APPC131091.1214 APPC131091.1611 APPC131091.1612
Methanol ≥99,9% (durch GC) Reag. Ph. Eur., ACS, ISO zur Analyse
Methanol
For analysis, ACS, and ISO.


Formel: CH₃OH
MW: 32.04 g/mol
Siedepunkt: 64...65 °C
Melting Pt: –97.8 °C
Dichte: 0.792 kg/l
Flash-Pt: 11
Storage Temperature: Raumtemperatur
MDL Number: MFCD00004595
CAS-Nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1) II
Merck Index: 13,05984
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=7bb8f9d74455604a6191a480ba063529ad0783c7&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Minimum Assay (G.C.) 99.9%
Identity IR Passes Test
Density 20/4 0.791 - 0.79
APHA colour 10
Acidity 0.0002 meq/g
Alkalinity 0.0002 meq/g
Insoluble Matter in H₂O Passes Test
Non-Volatile Matter 0.0005%
Reducing Substance to KMnO₄ (as O) 0.00025%
Darkened Substances by H₂SO₄ Passes Test
Carbonyl Compounds (as CH₃COCH₃) 0.002%
Acetaldehyde (CH₃CHO) 0.001%
Acetone (G.C.) 0.001%
Formaldehyde (HCHO) 0.001%
Ethanol (G.C.) 0.005%
2-Propanol (G.C.) 0.005%
Water(H₂O) 0.05%
Metals by ICP, Ag 0.05 mg/kg (ppm)
Metals by ICP, Al 0.5 mg/kg (ppm)
Metals by ICP, As 0.05 mg/kg (ppm)
Metals by ICP, Au 0.05 mg/kg (ppm)
Metals by ICP, B 0.02 mg/kg (ppm)
Metals by ICP, Ba 0.1 mg/kg (ppm)
Metals by ICP, Be 0.02 mg/kg (ppm)
Metals by ICP, Bi 0.05 mg/kg (ppm)
Metals by ICP, Ca 0.1 mg/kg (ppm)
Metals by ICP, Cd 0.05 mg/kg (ppm)
Metals by ICP, Co 0.02 mg/kg (ppm)
Metals by ICP, Cr 0.02 mg/kg (ppm)
Metals by ICP, Cu 0.02 mg/kg (ppm)
Metals by ICP, Fe 0.1 mg/kg (ppm)
Metals by ICP, Ga 0.02 mg/kg (ppm)
Metals by ICP, Ge 0.05 mg/kg (ppm)
Metals by ICP, Hg 0.05 mg/kg (ppm)
Metals by ICP, In 0.05 mg/kg (ppm)
Metals by ICP, K 0.1 mg/kg (ppm)
Metals by ICP, Li 0.05 mg/kg (ppm)
Metals by ICP, Mg 0.05 mg/kg (ppm)
Metals by ICP, Mn 0.02 mg/kg (ppm)
Metals by ICP, Mo 0.02 mg/kg (ppm)
Metals by ICP, Na 0.5 mg/kg (ppm)
Metals by ICP, Ni 0.02 mg/kg (ppm)
Metals by ICP, P 0.2 mg/kg (ppm)
Metals by ICP, Pb 0.02 mg/kg (ppm)
Metals by ICP, Pt 0.02 mg/kg (ppm)
Metals by ICP, S 0.2 mg/kg (ppm)
Metals by ICP, Sb 0.02 mg/kg (ppm)
Metals by ICP, Si 0.2 mg/kg (ppm)
Metals by ICP, Sn 0.1 mg/kg (ppm)
Metals by ICP, Sr 0.2 mg/kg (ppm)
Metals by ICP, Ti 0.02 mg/kg (ppm)
Metals by ICP, Tl 0.02 mg/kg (ppm)
Metals by ICP, V 0.02 mg/kg (ppm)
Metals by ICP, Zn 0.1 mg/kg (ppm)
Metals by ICP, Zr 0.02 mg/kg (ppm)

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge