Mineral oil light USP, NF, Ph. Eur., BP, pharma grade
Lieferant: APPLICHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
Gefahr
146257.1211
APPC146257.1211EA 45 EUR InStock
APPC146257.1211
Mineral oil light USP, NF, Ph. Eur., BP, pharma grade
Mineral oil light
Siedepunkt: 300...450 °C
Dichte: 0.845 g/cm³
Flash-Pt: >180 °C
MDL Number: MFCD00131611
CAS-Nummer: 8042-47-5
EINECS: 232-455-8
Order Now
Spezifikation Testergebnisse
Identity According to Pharmacopoeias Passes Test
Density 20/20 0.810 - 0.875
Density 25/25 0.818 - 0.880
Viscosity Kinematic (39.9 - 40.1 °C) 3.0 - 33.5 mm²/s
Viscosity Dynamic (19.9 - 20.1 °C) 25 - 80 mPas
Acidity or Alkalinity Passes Test
Darkened Substances by H₂SO₄ Passes Test
Sulphur Compounds Passes Test
Solid Paraffin Passes Test
Residual Solvents (Ph.Eur./USP) Passes Test
Polycyclic Aromatic Hydrocarbons Passes Test
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge