LDPE Tubing antistatic pink 150mm x 100 m Roll antistatic, definition: ultra clean, antistatic pink PE tubing Cleanliness level: 200 (according to IEST-STD-CC1246D) ASRE00-0231-0001 1 * 1 Rolle
Lieferant: PALL LIFESCIENCES
LDPE Tubing antistatic pink 150mm x 100 m Roll antistatic, definition: ultra clean, antistatic pink PE tubing Cleanliness level: 200 (according to IEST-STD-CC1246D) ASRE00-0231-0001 1 * 1 Rolle
PRODUKT
Erweitert / Kompakt
LDPE Tubing antistatic pink 150mm x 100 m Roll antistatic, definition: ultra clean, antistatic pink PE tubing Cleanliness level: 200 (according to IEST-STD-CC1246D) ASRE00-0231-0001 1 * 1 Rolle ATMIASRE0002310001 1 * 1 Rolle
Hersteller-Artikelnummer:: ASRE0002310001
Lokale Artikelnummer:: ATMIASRE0002310001
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder