Anti-H2AFJ Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ARP67131_P050
AVIVARP67131_P050EA 401 EUR InStock
AVIVARP67131_P050
Anti-H2AFJ Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: H2AFJ (H2A histone family, member J)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge