Mouse APOC1 ELISA Kit
Lieferant: AVIVA SYSTEMS BIOLOGY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
OKEH03072
AVIVOKEH03072EA 874 EUR InStock
AVIVOKEH03072
Mouse APOC1 ELISA Kit
Tests ELISAs
Aviva Systems Biology APOC1 ELISA Kit (Mouse) (OKEH03072) is based on standard sandwich enzyme-linked immuno-sorbent assay technology. An antibody specific for Apoc1 has been pre-coated onto a 96-well plate (12 x 8 Well Strips) and blocked. Standards or test samples are added to the wells and removed. A biotinylated detector antibody specific for Apoc1 is added, incubated and followed by washing. Avidin-Peroxidase Conjugate is then added, incubated and unbound conjugate is washed away. An enzymatic reaction is produced through the addition of TMB substrate which is catalyzed by HRP generating a blue color product that changes yellow after adding acidic stop solution. The density of yellow coloration read by absorbance at 450 nm is quantitatively proportional to the amount of sample Apoc1 captured in well.
  • Mean recovery when spiking into sample matrices at concentrations within the dynamic range: 102%
Mean Intra-assay CV%: <= 7.9% (n = 20)
Mean Inter-assay CV%: <= 10.1% (n = 20)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge