Robotic tips for Hamamatsu 6000 and 7000 systems, Axygen®
Lieferant: Corning
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
FDSS-1536-BK-R
AXYGFDSS-1536-BK-REA 9230 EUR InStock
AXYGFDSS-1536-BK-R
Robotic tips for Hamamatsu 6000 and 7000 systems, Axygen®
Pipettenspitzen
These black automation tips made of PP are engineered to the exact specifications of the original equipment manufacturers’ tips, ensuring complete compatibility and a consistent fit.
  • Conductive tips
  • Certified DNase-/RNase-free and non pyrogenic for complete protection against contamination
  • Optimised fine point for accurate dispensing
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge