Deep well plates, 24-well, Axygen®
Lieferant: Corning
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
P-DW10ML24C P-DW10ML24CS PDW10ML24C2BCS
AXYGP-DW10ML24CEA 226 EUR InStock
AXYGP-DW10ML24C AXYGP-DW10ML24CS AXYGPDW10ML24C2BCS
Deep well plates, 24-well, Axygen®
Mikrotiterplatten Deep-Well-Mikroplatten
Deep well and assay plates are used for sample collection, in vitro growth chambers and longterm storage. Made of PP, they offer excellent chemical resistance and will withstand temperatures down to –80 °C. Ultra-flat surface ensures proper sealing with heat sealing film. Clear V-shaped well bottom.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge