384-well Tips, 70 µl, SLAS rack, Axygen®
Lieferant: Corning
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VT-384-70-LRS
AXYGVT-384-70-LRSEA 5970 EUR InStock
AXYGVT-384-70-LRS
384-well Tips, 70 µl, SLAS rack, Axygen®
Pipettenspitzen
Axygen® offers a comprehensive line of tips for Agilent®/Velocity11 automation systems.
  • Maximum Recovery technology for ultra-low retention
  • RNase-/DNase-free and nonpyrogenic
  • Clear polypropylene tips
  • Non filtered
  • Sterile
  • Low Retention
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
Compatibility: Agilent®/Velocity VPrep® and Bravo 384ST, 96ST and 16ST head.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge