Polyethylene stoppers
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
TRF591 TRF592 TRF593 TRF595 TRF596 TRF597 TRF598
217-0716EA 27 EUR InStock
217-0716 217-0717 217-0718 AZLOTRF595 AZLOTRF596 AZLOTRF597 AZLOTRF598
Polyethylene stoppers
Stopfen PE/LDPE/HDPE-Stopfen
These PE stoppers are ideally suited for use with glass or plastic volumetric flasks and measuring cylinders etc. which have longer 'B' length sockets.
Large head provides a good grip and gives protection if glassware is accidently knocked over.
Verpackung: 10/package
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge