Safety wash bottles, wide neck, venting, multilingual
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
WGW537VTMLA WGW538VTMLA WGW539VTMLA WGW541VTMLA WGW601PMLA
AZLOWGW537VTMLAEA 55 EUR InStock
AZLOWGW537VTMLA AZLOWGW538VTMLA AZLOWGW539VTMLA AZLOWGW541VTMLA AZLOWGW601PMLA
Safety wash bottles, wide neck, venting, multilingual
Flaschen Spritzflaschen
LDPE wash bottles gives accurate dispensation of liquid.
  • The patented DripLok® vapour venting valve prevents solvent drips
  • Indelibly printed in three languages (English, French and Spanish)
  • Closure feature precision jet-tip aperture for fine control
Wide neck for easy, safe filling. Fitted with colour coded one-piece PP closure that maintain their shape in use.
Zertifizierungen: Wash bottles meet the material requirements of the U.S. Food and Drug Administration as specified in 21 CFR 177.1520.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge