Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Bathophenanthrolindisulfonsäure Dinatriumsalz Hydrat ≥95%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


M-Clarity™ Qualitätsstufe = MQ200

B1375-1G B1375-500MG B1375-5G
B1375-1GEA 253 EUR InStock
B1375-1G B1375-500MG B1375-5G
Bathophenanthrolindisulfonsäure Dinatriumsalz Hydrat ≥95%, Sigma-Aldrich®
Bathophenanthrolindisulfonsäure Dinatriumsalz Hydrat
Bathophenanthrolinedisulfonic acid disodium salt hydrate has been used to measure iron to determine the solubility of iron in the biological sample. It has been used as an iron chelator to grow plants without Fe-EDTA (ethylenediaminetetraacetic acid).

Bathophenanthrolinedisulfonic acid is a metal chelator. It permits iron determination to be made in aqueous medium without need for solvent extraction. It forms a complex with Fe2+ and makes it unavailable to Fe2+ transporter, thereby inhibiting the transport of iron into chloroplasts. Bathophenanthrolinedisulfonic acid is also used in the assay for determining intracellular glutathione.


Formel: C₂₄H₁₄N₂Na₂O₆S₂·H₂O
MW: 536.48 g/mol (anhy)
Melting Pt: 300 °C (Lit.)
Storage Temperature: Room Temperature
MDL Number: MFCD00149310
CAS-Nummer: 52746-49-3
EINECS: 258-152-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99351a037f8036de14f21f0408aef75cc4057bcd&vid=99351a037f8036de14f21f0408aef75cc4057bcd&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (colour) White to Orange and Faint Beige to Beige and Pink
Appearance (form) Powder
Titration (NT) HCLO₄ 0.1M calculated on dry substance
Purity (HPLC area %) ≥ 94.5 %
Solubility (turbidity) Clear to Slightly Hazy
Solubility (method) 100 mg/ml in water
Water Report Result

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge