B20054.06
B20054.14
B20054.06EA
97.8
EUR
InStock
B20054.06
B20054.14
trans-4-Methylcyclohexanol ≥98%
trans-4-Methylcyclohexanol
Purity: 98%
CAS No.: 7731-29-5
Molecular Formula: C7H14O
Formula Weight: 114.19
Flash Point: 72°(162°F)
Refractive Index: 1.4550
Boiling Point: 173-175°
MDL No.: MFCD00064171